missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Gluconate, USP, 97-102%, Spectrum™ Chemical
FE109, 12389-15-0, C12H22FeO14·2H2O
$442.63 - $9234.80
Chemical Identifiers
CAS | 12389-15-0 |
---|---|
Molecular Formula | C12H26FeO16 |
Molecular Weight (g/mol) | 482.17 |
InChI Key | OKGNXSFAYMSVNN-UHFFFAOYNA-L |
IUPAC Name | λ²-iron(2+) bis(2,3,4,5,6-pentahydroxyhexanoate) dihydrate |
SMILES | O.O.[Fe++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18602703
|
Spectrum Chemical Mfg Cor
FE109500GMPL |
500 g | Poly Bottle |
Each for $442.63
|
|
||||
18602704
|
Spectrum Chemical Mfg Cor
FE10925KGPL |
2.5 kg | Poly Pail |
Each for $1,330.30
|
|
||||
18602705
|
Spectrum Chemical Mfg Cor
FE10912KG |
12 kg | Poly Pail |
Each for $3,539.77
|
|
||||
18602706
|
Spectrum Chemical Mfg Cor
FE10950KGBL |
50 kg | Fiber Drum |
Each for $9,234.80
|
|
||||
Description
Spectrum™ Chemical Ferrous Gluconate, USP is used to treat iron deficiency anemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
12389-15-0 | |
482.17 | |
λ²-iron(2+) bis(2,3,4,5,6-pentahydroxyhexanoate) dihydrate |
C12H26FeO16 | |
OKGNXSFAYMSVNN-UHFFFAOYNA-L | |
O.O.[Fe++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O |
Specifications
12389-15-0 | |
500 g | |
OKGNXSFAYMSVNN-UHFFFAOYNA-L | |
λ²-iron(2+) bis(2,3,4,5,6-pentahydroxyhexanoate) dihydrate | |
97 to 102% | |
6.5-10% |
100% | |
C12H26FeO16 | |
O.O.[Fe++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O | |
482.17 | |
USP | |
Poly Bottle |