missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fenofibric acid

Chemical Identifiers

CAS 42017-89-0
Chemical Name or Material Fenofibric acid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466170010
View Documents
Thermo Scientific™
466170010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC466170050
View Documents
Thermo Scientific™
466170050
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

42017-89-0
Fenofibric acid

Specifications

42017-89-0
Solubility aq. soln.: .. Other solubilities: practically insoluble in chloroform, soluble in acetone and methanol
Fenofibric acid
1 g
187.0°C
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

Toxic to aquatic life with long lasting effects.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

EINECSNumber : 255-626-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only