Learn More
3M™ Faceseal L-120-5
Designed to be used with 3M™ Headgear L-500 and L-700 Series. This faceseal is used in applications with high temperatures or when sparks or molten metal may be present.
Supplier:  3M™ 7000126312
Catalog No. 29-636-6689
Only null left
Explore available promotions
Description
Description
  • For use with 3M™ Headgear L-Series
Specifications
Specifications
Face Seal
5/Cs.
Product Suggestions
Product Suggestions