Learn More
MilliporeSigma EZ-Fit™ Filtration Unit with Absorbent Pad
Membrane filtration device made of gridded, mixed cellulose esters membrane for bioburden testing
$359.05 - $374.05
Specifications
Disposable Single-use
Material Polystyrene
Sample Volume 100 mL
Sterility Sterile
Type Filtration Unit Without Absorbent Pad
View More Specs
Products 2
Catalog Number Mfr. No. Grid Color Price Quantity  
Catalog Number Mfr. No. Grid Color Price Quantity  
EFHAB100M3
View Documents
MilliporeSigma
EFHAB100M3
White
Pack of 48 Each for $374.05
Only null left
EFHAW100M3
View Documents
MilliporeSigma
EFHAW100M3
Blue
Pack of 48 Each for $359.05
Only null left
Description
Description
The EZ-FIT™ filtration unit is a single-use filtration device made of white, gridded, and mixed cellulose esters membrane with a blue base integrating a cellulose support pad.
  • Provides a safe and fast way of performing membrane filtration by avoiding the exposure to the risks of microbiological analysis
  • E-beam irradiated
  • Vented lid covers the sample throughout filtration
  • 360° rim prevents forceps from accidentally touching the membrane filtration area during membrane transfer
Specifications
Specifications
Single-use
100 mL
Filtration Unit Without Absorbent Pad
Polystyrene
Sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'