missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ External-Controlled Hot Plate, 150°C to 371°C, Porcelain-coated Stainless Steel
Enable uniform heating of large volumes and multiple vessels.
$4222.33 - $5327.87
Specifications
Top Plate Material | Porcelain-coated Stainless Steel |
---|---|
Temperature (Metric) | 150°C to 371°C |
Amperage | 13.3 A |
Frequency | 50/60 Hz |
Max. Sample Load | 18 kg |
Description
Enable uniform heating of large volumes and multiple vessels using Thermo Scientific™ External-Controlled Hotplates. These hotplates are ideal for a range of applications with acidic or basic aqueous solutions. An external controller operates outside fume hoods and corrosive environment.
Key Features:- Isolation of controls allows operation in hazardous environments
- Large 12 x 12″ (30.5 x 30.5 cm) and 12 x 24″ (30.5 x 60.9 cm) top plate surfaces accommodate up to 40 lb. (18.1 kg) loads
- Power light indicates when control is operating
- Large capacity with excellent temperature stabilityy
- Temperature stability is ±5°C (9°F) over the operating temperature range Embedded heating elements maximize temperature uniformity and evenly transfer heat within ±10°C (18°F) at 150°C (300°F)
- Percentage input controller delivers infinite temperature selection
Construction:
- Porcelain-coated, stainless-steel top offers chemical and stain resistance
- Chemical-resistant cover protects line cord
- Stainless-steel case offers maximum resistance to corrosion
Includes:
5′ (1.5 m) 3-wire cord and plug
Specifications
Porcelain-coated Stainless Steel | |
13.3 A | |
18 kg | |
Hot Plate |
150°C to 371°C | |
50/60 Hz | |
Rectangle | |
12 in. x 24 in. |