Learn More
Europium rod, 9.52 mm (0.375in) dia, 99.9% (metals basis)
CAS: 7440-53-1 | Eu | 151.96 g/mol
$792.62 - $3230.51
Chemical Identifiers
CAS | 7440-53-1 |
---|---|
Molecular Formula | Eu |
Molecular Weight (g/mol) | 151.96 |
MDL Number | MFCD00010992 |
InChI Key | OGPBJKLSAFTDLK-UHFFFAOYSA-N |
Synonym | europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil |
PubChem CID | 23981 |
ChEBI | CHEBI:32999 |
IUPAC Name | europium |
SMILES | [Eu] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged in mineral oil
Chemical Identifiers
7440-53-1 | |
151.96 | |
OGPBJKLSAFTDLK-UHFFFAOYSA-N | |
23981 | |
europium |
Eu | |
MFCD00010992 | |
europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil | |
CHEBI:32999 | |
[Eu] |
Specifications
Gray-Silver | |
10 mm | |
≈0.37g/mm | |
Eu | |
UN3208 | |
OGPBJKLSAFTDLK-UHFFFAOYSA-N | |
europium | |
23981 | |
99.9% | |
(metals basis) |
7440-53-1 | |
Odorless | |
Cast Rod | |
MFCD00010992 | |
europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil | |
[Eu] | |
151.96 | |
CHEBI:32999 | |
Europium rod, ≈9.52 mm (0.375 in.) dia. |
Safety and Handling
GHS H Statement
H250-H261
P210-P222-P223-P231+P232-P280-P335+P334-P370+P378q-P501c
H250-H261-H500
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.
EINECSNumber : 231-161-7
TSCA : Yes
storageNote1 :
- Packaged in mineral oil
Recommended Storage : Ambient temperatures
RUO – Research Use Only