missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eugenol, USP, Spectrum™ Chemical
EU110, 97-53-0, C10H12O2
$274.17 - $79965.26
Chemical Identifiers
CAS | 97-53-0 |
---|---|
Molecular Formula | C10H12O2 |
Molecular Weight (g/mol) | 164.20 |
MDL Number | MFCD00008654 |
InChI Key | RRAFCDWBNXTKKO-UHFFFAOYSA-N |
IUPAC Name | 2-methoxy-4-(prop-2-en-1-yl)phenol |
SMILES | COC1=CC(CC=C)=CC=C1O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18602592
|
Spectrum Chemical Mfg Cor
EU110100ML |
100 mL | Amber Glass Bottle |
Each for $274.17
|
|
||||
18602593
|
Spectrum Chemical Mfg Cor
EU110500MLGL |
500 mL | Amber Glass Bottle |
Each for $680.97
|
|
||||
18602594
|
Spectrum Chemical Mfg Cor
EU1104LTGL |
4 L | Amber Glass Bottle |
Each for $2,846.34
|
|
||||
18602595
|
Spectrum Chemical Mfg Cor
EU11020LTBL |
20 L | Amber Glass Bottle |
Each for $8,223.23
|
|
||||
18602596
|
Spectrum Chemical Mfg Cor
EU110200LTBL |
200 L | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Eugenol, USP is used as a local antiseptic and anaesthetic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
97-53-0 | |
164.20 | |
RRAFCDWBNXTKKO-UHFFFAOYSA-N | |
COC1=CC(CC=C)=CC=C1O |
C10H12O2 | |
MFCD00008654 | |
2-methoxy-4-(prop-2-en-1-yl)phenol |
Specifications
97-53-0 | |
253°C | |
C10H12O2 | |
MFCD00008654 | |
RRAFCDWBNXTKKO-UHFFFAOYSA-N | |
2-methoxy-4-(prop-2-en-1-yl)phenol | |
USP |
100% | |
Amber Glass Bottle | |
1.540-1.542 | |
100 mL | |
COC1=CC(CC=C)=CC=C1O | |
164.20 | |
Liquid |