missing translation for 'onlineSavingsMsg'
Learn More

Etravirine, Thermo Scientific™

$236.40 - $595.66

Chemical Identifiers

CAS 269055-15-4
Molecular Formula C20H15BrN6O
Molecular Weight (g/mol) 435.29
MDL Number MFCD09837879
InChI Key PYGWGZALEOIKDF-UHFFFAOYSA-N
Synonym 4-((6-Amino-5-bromo-2-((4-cyanophenyl)amino)pyrimidin-4-yl)oxy)-3,5-dimethylbenzonitrile
IUPAC Name 4-({6-amino-5-bromo-2-[(4-cyanophenyl)amino]pyrimidin-4-yl}oxy)-3,5-dimethylbenzonitrile
SMILES CC1=CC(=CC(C)=C1OC1=NC(NC2=CC=C(C=C2)C#N)=NC(N)=C1Br)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC468721000
View Documents
Thermo Scientific Chemicals
468721000
100 mg
Each for $236.40
Only null left
 
AC468725000
View Documents
Thermo Scientific Chemicals
468725000
500 mg
Each for $595.66
Only null left
 
Description

Description

Specifications

Chemical Identifiers

269055-15-4
435.29
PYGWGZALEOIKDF-UHFFFAOYSA-N
4-({6-amino-5-bromo-2-[(4-cyanophenyl)amino]pyrimidin-4-yl}oxy)-3,5-dimethylbenzonitrile
C20H15BrN6O
MFCD09837879
4-((6-Amino-5-bromo-2-((4-cyanophenyl)amino)pyrimidin-4-yl)oxy)-3,5-dimethylbenzonitrile
CC1=CC(=CC(C)=C1OC1=NC(NC2=CC=C(C=C2)C#N)=NC(N)=C1Br)C#N

Specifications

Glass Bottle
269055-15-4
C20H15BrN6O
White to Yellow to Beige
4-((6-Amino-5-bromo-2-((4-cyanophenyl)amino)pyrimidin-4-yl)oxy)-3,5-dimethylbenzonitrile
PYGWGZALEOIKDF-UHFFFAOYSA-N
4-({6-amino-5-bromo-2-[(4-cyanophenyl)amino]pyrimidin-4-yl}oxy)-3,5-dimethylbenzonitrile
100 mg
Etravirine
Available
Powder
MFCD09837879
262°C to 265°C
Insoluble in water
CC1=CC(=CC(C)=C1OC1=NC(NC2=CC=C(C=C2)C#N)=NC(N)=C1Br)C#N
435.29
435.28
Videos
Safety and Handling

Safety and Handling

GHS07: Exclamation mark

ShelfLife : 5 years

Recommended Storage : Freezer -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'