missing translation for 'onlineSavingsMsg'
Learn More

Etoposide 98.0+%, TCI America™

$205.05 - $822.05

Chemical Identifiers

CAS 33419-42-0
Molecular Formula C29H32O13
Molecular Weight (g/mol) 588.56
MDL Number MFCD00869325,MFCD00869325
InChI Key VJJPUSNTGOMMGY-MRVIYFEKSA-N
Synonym vjjpusntgommgy-nzlmilqcsa
PubChem CID 50936917
IUPAC Name (10R,11R,15R,16S)-16-{[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
SMILES COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E0675100MG
View Documents
TCI America
E0675100MG
100 mg
Each for $205.05
Only null left
 
E06751G
View Documents
TCI America
E06751G
1 g
Each for $822.05
Only null left
 
Description

Description

Specifications

Chemical Identifiers

33419-42-0
588.56
VJJPUSNTGOMMGY-MRVIYFEKSA-N
50936917
COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12
C29H32O13
MFCD00869325,MFCD00869325
vjjpusntgommgy-nzlmilqcsa
(10R,11R,15R,16S)-16-{[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one

Specifications

33419-42-0
White
MFCD00869325,MFCD00869325
vjjpusntgommgy-nzlmilqcsa
COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12
588.56
588.56
Crystalline Powder
251°C
C29H32O13
100 mg
VJJPUSNTGOMMGY-MRVIYFEKSA-N
(10R,11R,15R,16S)-16-{[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
50936917
≥98.0% (HPLC)
Etoposide
Videos
Safety and Handling

Safety and Handling

RTECSNumber : KC0190000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'