missing translation for 'onlineSavingsMsg'
Learn More

Etoposide, MP Biomedicals

$181.80 - $181.80

Chemical Identifiers

CAS 33419-42-0
Molecular Formula C29H32O13
Molecular Weight (g/mol) 588.56
MDL Number MFCD00869325,MFCD00869325
InChI Key VJJPUSNTGOMMGY-MRVIYFEKSA-N
Synonym vjjpusntgommgy-nzlmilqcsa
PubChem CID 50936917
SMILES COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19391825
View Documents
MP Biomedicals Inc
0219391825
25 mg
Each for $181.80
Only null left
 
Description

Description

Specifications

Chemical Identifiers

33419-42-0
588.56
VJJPUSNTGOMMGY-MRVIYFEKSA-N
50936917
C29H32O13
MFCD00869325,MFCD00869325
vjjpusntgommgy-nzlmilqcsa
COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12

Specifications

33419-42-0
C29H32O13
25 mg
VJJPUSNTGOMMGY-MRVIYFEKSA-N
(10R,11R,15R,16S)-16-{[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
50936917
Powder
White
MFCD00869325,MFCD00869325
vjjpusntgommgy-nzlmilqcsa
COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12
588.56
588.6
Etoposide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 251-509-1

Recommended Storage : Store at Room Temperature (15 to 30°C).

SDS
Documents

Documents