missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediamine Dihydroiodide 98.0+%, TCI America™
$126.62 - $385.51
Chemical Identifiers
CAS | 5700-49-2 |
---|---|
Molecular Formula | C2H10I2N2 |
Molecular Weight (g/mol) | 315.925 |
MDL Number | MFCD00035535 |
InChI Key | IWNWLPUNKAYUAW-UHFFFAOYSA-N |
Synonym | ethylenediamine dihydroiodide, jodethamine, ethane-1,2-diamine dihydroiodide, hydrodine, eddi, ethanediamine dihydroiodide, ethylenediammonium diiodide, unii-721m52nq5l, ethylenediamine dihydroiodide, 1,2-ethanediamine dihydriodide |
PubChem CID | 21921 |
ChEBI | CHEBI:81719 |
IUPAC Name | ethane-1,2-diamine;dihydroiodide |
SMILES | C(CN)N.I.I |
Chemical Identifiers
5700-49-2 | |
315.925 | |
IWNWLPUNKAYUAW-UHFFFAOYSA-N | |
21921 | |
ethane-1,2-diamine;dihydroiodide |
C2H10I2N2 | |
MFCD00035535 | |
ethylenediamine dihydroiodide, jodethamine, ethane-1,2-diamine dihydroiodide, hydrodine, eddi, ethanediamine dihydroiodide, ethylenediammonium diiodide, unii-721m52nq5l, ethylenediamine dihydroiodide, 1,2-ethanediamine dihydriodide | |
CHEBI:81719 | |
C(CN)N.I.I |
Specifications
5700-49-2 | |
C2H10I2N2 | |
1 g | |
IWNWLPUNKAYUAW-UHFFFAOYSA-N | |
ethane-1,2-diamine;dihydroiodide | |
21921 | |
315.92 | |
Crystalline Powder |
White-Yellow | |
MFCD00035535 | |
ethylenediamine dihydroiodide, jodethamine, ethane-1,2-diamine dihydroiodide, hydrodine, eddi, ethanediamine dihydroiodide, ethylenediammonium diiodide, unii-721m52nq5l, ethylenediamine dihydroiodide, 1,2-ethanediamine dihydriodide | |
C(CN)N.I.I | |
315.925 | |
CHEBI:81719 | |
≥98.0% (N,T) | |
Ethylenediamine Dihydroiodide |
Safety and Handling
TSCA : Yes