Learn More
Ethanolamine, ACS reagent
CAS: 141-43-5 | C2H7NO | 61.08 g/mol
$55.85 - $306.83
Chemical Identifiers
CAS | 141-43-5 |
---|---|
Molecular Formula | C2H7NO |
Molecular Weight (g/mol) | 61.08 |
MDL Number | MFCD00008183 |
InChI Key | HZAXFHJVJLSVMW-UHFFFAOYSA-N |
Synonym | ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine |
PubChem CID | 700 |
ChEBI | CHEBI:16000 |
SMILES | NCCO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC451762500
|
Thermo Scientific Chemicals
451762500 |
250 mL |
Each for $55.85
|
|
|||||
AC451760010
|
Thermo Scientific Chemicals
451760010 |
1 L |
Each for $147.53
|
|
|||||
AC451760025
|
Thermo Scientific Chemicals
451760025 |
2.5 L |
Each for $306.83
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141-43-5 | |
61.08 | |
HZAXFHJVJLSVMW-UHFFFAOYSA-N | |
700 | |
NCCO |
C2H7NO | |
MFCD00008183 | |
ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine | |
CHEBI:16000 |
Specifications
141-43-5 | |
1.0120g/mL | |
92°C | |
C2H7NO | |
H2NCH2CH2OH | |
250 mL | |
15,3782 | |
Solubility in water: soluble. Other solubilities: miscible with methanol and acetone, soluble in benzene, ether, ccl4 and n-heptane | |
NCCO | |
61.08 | |
24 mPa.s (20°C) | |
61.08 | |
ACS Reagent |
10.3°C | |
170.0°C | |
Authentic | |
1.4530 to 1.4550 | |
MFCD00008183 | |
1.012 | |
ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine | |
HZAXFHJVJLSVMW-UHFFFAOYSA-N | |
2-aminoethanol | |
700 | |
CHEBI:16000 | |
≥99.0% (GC) | |
Ethanolamine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:
GHS Signal Word: Danger
EINECSNumber : 205-483-3
RUO – Research Use Only