missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NoTrax™ Ergo Trax™ Grande™
Ergo Trax™ Grande™ a thicker, heavier and stronger anti-fatigue floor mat
Supplier: NoTrax™ 985S0035YB
Description
- Thicker, heavier, stronger anti-fatigue floor mat
- For industrial applications and multi-shift operations in dry work areas
- Thickness: 1 in.
- Maximizes worker productivity and comfort
- Durable vinyl diamond-plate top surface design provides non-directional traction
- Easy to clean
- Top surface combined with a Niru™ rubber sponge base utilizing NoTrax™ exclusive UniFusion™ technology
- Eliminates possibility of de-lamination
- Sloped beveled edges reduce tripping hazards and allows easy access on and off mat
Specifications
3 x 5 ft. | |
Black, Yellow | |
Anti-fatigue Floor Mat |
3 ft. | |
5 ft. | |
Heavy duty vinyl top surface combined with Niru™ rubber base |