Learn More
Agilent BioTek Epoch Microplate Spectrophotometer, IVD
Epoch microplate spectrophotometer has monochromator-based optics, for a filter-free, wide wavelength range for UV-Vis absorbance measurements. For In Vitro Diagnostic Use.
Supplier:  Agilent BioTek EPOCHRSI
 View more versions of this product
Catalog No. BTEPOCHRSI
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Epoch microplate spectrophotometer has monochromator-based optics, for a filter-free, wide wavelength range for UV-Vis absorbance measurements. These measurements can be done in many microplate formats and in 2 μL samples, when the available Take3 microvolume plate is used
Epoch is controlled with the Gen5 software interface, with simple programming and powerful data analysis. This robust, low maintenance microplate absorbance reader is a cost-effective system. Reliability and ease of use ensure time savings in laboratory workflows.
Specifications
Specifications
Microplate Reader
6 to 384-well Plates
±2 nm
CE and TUV marked. RoHS compliant.
12.5 in.
7.7 in.
50/60 Hz
EPOCHR
0.0001 OD
200 to 999 nm
6.8 kg
30.5 cm
Xenon Flash Lamp
200 to 999 nm, 1 nm increments
5 nm
0.5 mm
31.8 cm
19.6 cm
Epoch R Microplate Spectrophotometer; Gen5 microplate data analysis software
±0.2 nm
100/240 V
<15 lb.
12 in.
Product Suggestions
Product Suggestions