missing translation for 'onlineSavingsMsg'
Learn More

Epinastine Hydrochloride 98.0+%, TCI America™

$121.16 - $669.19

Chemical Identifiers

CAS 108929-04-0
Molecular Formula C16H16ClN3
Molecular Weight (g/mol) 285.78
MDL Number MFCD00933434
InChI Key VKXSGUIOOQPGAF-UHFFFAOYNA-N
Synonym 3-Amino-9,13b-dihydro-1H-dibenzo[c,f]imidazo[1,5-a]azepine Hydrochloride
PubChem CID 157313
ChEBI CHEBI:51037
IUPAC Name hydrogen 2,4-diazatetracyclo[12.4.0.0²,⁶.0⁷,¹²]octadeca-1(18),3,7,9,11,14,16-heptaen-3-amine chloride
SMILES [H+].[Cl-].NC1=NCC2N1C1=CC=CC=C1CC1=CC=CC=C21
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E0799100MG
View Documents
TCI America
E0799100MG
100 mg
Each for $121.16
Only null left
 
E07991G
View Documents
TCI America
E07991G
1 g
Each for $669.19
Only null left
 
Description

Description

Specifications

Chemical Identifiers

108929-04-0
285.78
VKXSGUIOOQPGAF-UHFFFAOYNA-N
157313
hydrogen 2,4-diazatetracyclo[12.4.0.0²,⁶.0⁷,¹²]octadeca-1(18),3,7,9,11,14,16-heptaen-3-amine chloride
C16H16ClN3
MFCD00933434
3-Amino-9,13b-dihydro-1H-dibenzo[c,f]imidazo[1,5-a]azepine Hydrochloride
CHEBI:51037
[H+].[Cl-].NC1=NCC2N1C1=CC=CC=C1CC1=CC=CC=C21

Specifications

108929-04-0
White
MFCD00933434
3-Amino-9,13b-dihydro-1H-dibenzo[c,f]imidazo[1,5-a]azepine Hydrochloride
[H+].[Cl-].NC1=NCC2N1C1=CC=CC=C1CC1=CC=CC=C21
285.78
CHEBI:51037
≥98.0% (HPLC,N)
Epinastine Hydrochloride
275°C
C16H16ClN3
100 mg
VKXSGUIOOQPGAF-UHFFFAOYNA-N
hydrogen 2,4-diazatetracyclo[12.4.0.0²,⁶.0⁷,¹²]octadeca-1(18),3,7,9,11,14,16-heptaen-3-amine chloride
157313
285.78
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'