Learn More
MilliporeSigma™ Supelco™ Empty Stainless Steel GC Column
For less demanding applications where the inertness of glass is not required, choose more economical stainless steel. Column can be carefully bent to fit most instruments.
$108.70 - $108.70
Specifications
Diameter (Metric) Inner | 2.1 mm |
---|---|
Type | Empty GC |
For Use With (Application) | Gas Chromatography (GC) |
Material | Stainless Steel Column |
Packing Material | None |
Description
Empty column may be used for packing adsorbents for temperature-programmed desorption (TPD) instrumentation.Specifications
2.1 mm | |
Gas Chromatography (GC) | |
None |
Empty GC | |
Stainless Steel Column |