Learn More
Thermo Scientific Chemicals Efaproxiral sodium, 98%
CAS: 170787-99-2 | C20H22NNaO4 | 363.38 g/mol
$715.74 - $715.74
Chemical Identifiers
CAS | 170787-99-2 |
---|---|
Molecular Formula | C20H22NNaO4 |
Molecular Weight (g/mol) | 363.38 |
InChI Key | SWDPIHPGORBMFR-UHFFFAOYSA-M |
Synonym | efaproxiral sodium, sodium 2-4-2-3,5-dimethylphenyl amino-2-oxoethyl phenoxy-2-methylpropanoate, rsr 13 sodium, unii-3l83qp52xi, rsr13 sodium, efaproxiral sodium usan, rsr-13 |
PubChem CID | 2725048 |
IUPAC Name | sodium;2-[4-[2-(3,5-dimethylanilino)-2-oxoethyl]phenoxy]-2-methylpropanoate |
SMILES | CC1=CC(=CC(=C1)NC(=O)CC2=CC=C(C=C2)OC(C)(C)C(=O)[O-])C.[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC460100010
|
Thermo Scientific Chemicals
460100010 |
1 g |
Each for $715.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
170787-99-2 | |
363.38 | |
efaproxiral sodium, sodium 2-4-2-3,5-dimethylphenyl amino-2-oxoethyl phenoxy-2-methylpropanoate, rsr 13 sodium, unii-3l83qp52xi, rsr13 sodium, efaproxiral sodium usan, rsr-13 | |
sodium;2-[4-[2-(3,5-dimethylanilino)-2-oxoethyl]phenoxy]-2-methylpropanoate |
C20H22NNaO4 | |
SWDPIHPGORBMFR-UHFFFAOYSA-M | |
2725048 | |
CC1=CC(=CC(=C1)NC(=O)CC2=CC=C(C=C2)OC(C)(C)C(=O)[O-])C.[Na+] |
Specifications
170787-99-2 | |
Authentic | |
Glass Bottle | |
1 g | |
SWDPIHPGORBMFR-UHFFFAOYSA-M | |
sodium;2-[4-[2-(3,5-dimethylanilino)-2-oxoethyl]phenoxy]-2-methylpropanoate | |
2725048 | |
98% |
13% max. | |
97.5% min. (HPLC) | |
C20H22NNaO4 | |
efaproxiral sodium, sodium 2-4-2-3,5-dimethylphenyl amino-2-oxoethyl phenoxy-2-methylpropanoate, rsr 13 sodium, unii-3l83qp52xi, rsr13 sodium, efaproxiral sodium usan, rsr-13 | |
CC1=CC(=CC(=C1)NC(=O)CC2=CC=C(C=C2)OC(C)(C)C(=O)[O-])C.[Na+] | |
363.38 | |
363.38 | |
Efaproxiral sodium |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
RUO – Research Use Only