missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA (Dihydrate), ACS Reagent (Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), Reagents
EDTA (Dihydrate), ACS Reagent (Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C120110050F4
Specifications
EDTA (Dihydrate), ACS Reagent (Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate) | |
252°C | |
4 to 6 | |
Fiber Drum | |
50 kg | |
Edetate Disodium, Disodium EDTA, (Ethylenedinitrilo)tetraacetic Acid disodium salt dihydrate | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
372.24 | |
Solid |
6381-92-6 | |
White | |
>252°C | |
C10H18N2Na2O10 | |
0.86 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
ACS |
Chemical Identifiers
6381-92-6 | |
372.24 | |
Edetate Disodium, Disodium EDTA, (Ethylenedinitrilo)tetraacetic Acid disodium salt dihydrate | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
C10H18N2Na2O10 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |