missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Howard Leight™ HL400 Ear Plugs Dispenser with Prefilled Canister

Howard Leight™ HL400 ear plug dispensers are used with disposable ear plugs to provide a convenient dispensing point while keeping ear plugs clean and readily available. They also keep ear plugs organized and contained to minimize product loss or waste. Dispenser can be mounted to a wall or placed on a flat surface.

$142.54 - $142.54

Specifications

ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Ear Plug Accessibilty to Workers and Facility Visitors in Reception Areas and Noisy Environments
High Visibility Yes
View More Specs

Products 1
Catalog Number Mfr. No. Product Type Noise Reduction Rating (NRR) Price Quantity  
Catalog Number Mfr. No. Product Type Noise Reduction Rating (NRR) Price Quantity  
19-085-634
View Documents
Honeywell™
HL400LLINTRO
HL400 dispenser with 1 prefilled canister of Laser Lite™ ear plugs 32 dB
Case of 400 Each for $142.54
Only null left
 
Description

Description

  • Dispensing point that protects ear plugs from dust while keeping them readily available for use
  • Makes ear plugs accessible to workers and facility visitors to remind them to protect their hearing
  • Dispense one or two at a time to minimize waste and supply clean ear plugs to each user
  • Position in reception areas and at entries to noisy work zones for easy access
  • includes dispenser and prefilled canister of ear plugs
  • Can be mounted to a wall or placed on a flat surface
Specifications

Specifications

Yes
Universal
Yes
400 Pairs/Dispenser
Uncorded
Single-use
Flat Surface, Wall Mountable
Class AL
Ear Plug Accessibilty to Workers and Facility Visitors in Reception Areas and Noisy Environments
Dispenser
Foam
Bell
No
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'