missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Square Laboratory Bottles with DIN Thread

Ergonomic handling due to angular shape, highly stable, good stackability. Alongside proven DURAN™ properties, a space saving of 44 % in comparison with standard laboratory bottles. Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working.

$392.08 - $781.08

Specifications

Material Glass
Quantity 10/Pk.
Product Type Square Laboratory Bottle
View More Specs

Products 4
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
09-841-285
View Documents
DWK Life Sciences
218202453
100 mL
Case of 10 Each for $392.08
Only null left
 
09-841-286
View Documents
DWK Life Sciences
218203655
250 mL
Case of 10 Each for $442.55
Only null left
 
09-841-287
View Documents
DWK Life Sciences
218204454
500 mL
Case of 10 Each for $550.55
Only null left
 
09-841-288
View Documents
DWK Life Sciences
218205459
1 L
Case of 10 Each for $781.08
Only null left
 
Description

Description

  • Typical applications: space-saving storage, space-saving transport
  • With screw-cap and pouring ring from PP (blue)
  • Service temperature limit of screw cap and pouring ring: +140 °C
  • Easy-to-read scale and large labelling field for easy marking, in fired-on, highly durable white ceramic
Specifications

Specifications

Glass
10/Pk.
Square Laboratory Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'