Learn More
DWK Life Sciences DURAN™ Premium Bottle, Clear, with DIN 168-1 Thread, GL 45, Graduated
Glass laboratory bottles with superior thermal and chemical resistance.
$1524.21 - $1717.03
Specifications
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage and transport of media and reagents; Shipping |
---|---|
Thread Size (Metric) | GL 45 |
Autoclavable | Autoclavable |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
02-900-128
|
DWK Life Sciences
1127077 |
500 mL |
Case of 10 Each for $1,717.03
|
|
|||||
02-900-127
|
DWK Life Sciences
1127076 |
250 mL |
Case of 10 Each for $1,586.68
|
|
|||||
02-900-126
|
DWK Life Sciences
1127075 |
100 mL |
Case of 10 Each for $1,524.21
|
|
|||||
Description
The DURAN™ premium GL 45 laboratory bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. The premium bottles are ideal for high temperature applications: the thermal shock resistance of the glass bottle is certified for a 160°C differential, and is combined with a high temperature fluoropolymer (PFA) screw cap and pouring ring that are suitable for use up to 200°C. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature a double graduated scale for filling volume and a reversed scale for pouring. The marking field and Retrace Code for manufacturing lot traceability are printed in permanent white enamel.
- Additional graduation marks and an opposite scaling (accurate to ± 5 %) simplify the reading of the filling level as well as the quantity withdrawn
- Colourant free product
- Reliable sealing due to PTFE-coated cap liner
- TUÌV tested thermal shock resistance of 160 K
- Complete with PFA pouring ring and cap
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
Specifications
Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage and transport of media and reagents; Shipping | |
GL 45 | |
Autoclavable |