missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Series 254 Coveralls, Clean-Processed and Sterile
Lightweight, breathable, and clean processed coveralls for use in ISO 4-5 Class 10-100 critical environments.
$695.17 - $851.77
Specifications
Product Type | Cleanroom Coveralls |
---|---|
Material | Tyvek™ Flashspun Polyethylene |
Color | White |
Disposable | Single-use |
Ankle Style | Elastic |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-985-404
|
DuPont™
IC254BWH2X0025CS |
2X-Large |
Case of 25 Each for $743.99
|
|
|||||
17-985-405
|
DuPont™
IC254BWH3X0025CS |
3X-Large |
Case of 25 Each for $788.65
|
|
|||||
17-985-401
|
DuPont™
IC254BWHMD0025CS |
Medium |
Case of 25 Each for $701.89
|
|
|||||
17985403
|
DuPont™
IC254BWHXL0025CS |
X-Large |
Case of 25 Each for $695.17
|
|
|||||
17-985-406
|
DuPont™
IC254BWH4X0025CS |
4X-Large |
Case of 25 Each for $851.77
|
|
|||||
17-985-400
|
DuPont™
MC254BWHSM0025CS |
Small |
Case of 25 Each for $701.89
|
|
|||||
17985402
|
DuPont™
IC254BWHLG0025CS |
Large |
Case of 25 Each for $701.89
|
|
|||||
Description
For use in controlled environments with high standards for particle and microbiological contamination control (pharmaceutical, biotech, medical device and electronic settings).
- Material and bound seams offer an excellent barrier while maintaining comfort and durability
- Gamma sterilized and clean processed
- Spun polyethylene creates barrier to particles, micro-organisms and non-hazardous liquids
- Lowest level particle shedding
- Bound neck and seams
- Elastic wrists and ankles
- Dolman sleeves
- Zipper closure
Should not be worn around open flame, heat source, sparks or ignition sources
Specifications
Cleanroom Coveralls | |
White | |
Elastic | |
Collared | |
Bound | |
Anti-static, Abrasion | |
Class 10-100 (ISO 4-5) |
Tyvek™ Flashspun Polyethylene | |
Single-use | |
Elastic | |
Zipper | |
Sterile | |
Yes |