missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Series 180 Coveralls, Attached Hood
Light and durable coveralls provide a barrier to particles, microorganisms, and non-hazardous light liquid splash.
$273.88 - $307.75
Specifications
Product Type | Cleanroom Coveralls |
---|---|
Material | Tyvek™ |
Color | White |
Disposable | Single-use |
Ankle Style | Elastic |
Alert:
Should not be worn around open flame, heat source, sparks or ignition sources.
Catalog Number | Mfr. No. | Size | Size (Metric) Chest | Length (Metric) Sleeve | Size (Metric) Inseam | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Size (Metric) Chest | Length (Metric) Sleeve | Size (Metric) Inseam | Price | Quantity | ||
17-978-123
|
DuPont™
IC180SWH2X002500 |
2X-Large | 109.2 to 116.8 cm | 97.1 cm | 77.47 cm |
Case of 25 Each for $290.28
|
|
||
17-978-124
|
DuPont™
IC180SWH3X002500 |
3X-Large | 116.8 to 132 cm | 97.7 cm | 80 cm |
Case of 25 Each for $307.75
|
|
||
17-978-120
|
DuPont™
IC180SWHMD002500 |
Medium | 91.4 to 96.5 cm | 84.4 cm |
Case of 25 Each for $273.88
|
|
|||
17-978-121
|
DuPont™
IC180SWHLG002500 |
Large | 96.5 to 101.6 cm | 88.9 cm |
Case of 25 Each for $273.88
|
|
|||
17-978-122
|
DuPont™
IC180SWHXL002500 |
X-Large | 101.6 to 109.2 cm | 92.7 cm | 74.9 cm |
Case of 25 Each for $273.88
|
|
||
Description
IsoClean protects against micro-organisms and particulates.
- Spinning process creates barrier to particles, micro-organisms, and nonhazardous liquids
- Lowest level particle shedding
- Recyclable
- Serged seams
- Standard hood
- Elastic wrists and ankles
- Zipper closure
- Use in controlled environments with high standards for particle and microbiological contamination control
- Pharmaceutical, biotech, medical device, electronic settings
Specifications
Cleanroom Coveralls | |
White | |
Elastic | |
Serged | |
Tears | |
Bulk |
Tyvek™ | |
Single-use | |
Elastic | |
Non-sterile | |
Yes | |
Class 1000-100,000 (ISO 6-8) |