Learn More
DuPont™ Tychem™ SL 122 Series Coveralls, Berry Amendment Compliant
Delivers effective protection against a range of chemical environments.
Supplier: DuPont™ SL122TWH2X0006BN
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Tychem SL, utilizes Saranex™ 23-P film laminated to Tyvek fabric to provide a rugged and durable fabric that resists abrasion.
The fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.
- Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
- Attached hood with elastic around face opening
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Integrated socks composed of garment material
Specifications
Hooded Coveralls with Socks | |
Tyvek with Saranex 23-P Film Laminate | |
Single-use | |
Elastic | |
Attached, Elastic Face | |
Taped | |
Attached hood, elastic wrist, attached socks | |
30 in. | |
12 mil | |
38.25 in. | |
28.25 in. | |
6.08 to 6.33 ft. |
2X-Large | |
White | |
Attached Socks | |
Hooded | |
Front Zipper | |
Non-sterile | |
44.25 to 47 .75 in. | |
76.2 cm | |
Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments | |
97.15 cm | |
71.75 cm | |
1.82 to 1.92 m |