Learn More
DuPont™ ProShield™ 60 Coveralls, Respirator Fit Hood and Storm Flap
Coveralls with respirator fit hood are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
Supplier:  DuPont™ NG127SWHMD0025NP
 View more versions of this product
Catalog No. 29-634-247
Only null left
Explore available promotions
Description
Description
  • Microporous film that is laminated to a spundbonded polypropylene substrate.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements.
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask.
  • Longer zipper extends to chin for complete coverage of neck area
  • Storm flap of garment material covers zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex.
  1. Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
  2. ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
  3. Certified incinerable by independent laboratory. For details, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Hooded
Front Zipper
Non-sterile
ASTM D751, D1117, D5034, D257, AATCC 127
Yes
33.75 to 37.25 in.
85.72 to 94.61 cm
10 mil
31.75 in.
5.3 to 5.7 ft.
25/Cs.
Medium
Proprietary Bilaminate Composite Fabric
Single-use
Elastic
Respirator Fit
Serged
Particle, Liquid
Microporous Film, Laminated to a Spundbonded Polypropylene Substrate
NG127S
26 in.
66.04 cm
Automotive Refinishing, Waste Cleanup, Asbestos Abatement, Hazardous Remediation
80.64 cm
1.6 to 1.73 m
Product Suggestions
Product Suggestions