Learn More
Dräger™ Haz Mat Simultest Test Kit
Simultaneously detects multiple contaminants
Supplier: Dräger™ 4056098
Alert:
No other type of tube should be substituted for use with these kits.
Includes:
- accuro™ pump
- Pump spare parts kit
- Air currents kit
- 39 in. (1m) extension hose with adapter (includes scoring tool and manifold)
- Simultaneous test set opener
- Deluxe tube opener
- Laminated instruction cards
- Three each of Simultaneous Test Set† I (tests for hydrogen chloride, hydrogen cyanide, carbon monoxide, ammonia, and nitrous fumes)
- Three each of Simultaneous Test Set† II (tests for sulfur dioxide, chlorine, hydrogen sulfide, carbon dioxide, and phosgene)
- Four each of Simultaneous Test Set† III (tests for alcohols, aliphatic hydrocarbons, aromatics, chlorinated hydrocarbons, and ketones)
- Hard-plastic carrying case
Description
Provides Hazmat teams with onsite information about levels of inorganic/organic gases and vapors encountered during fires and afterward during salvage and overhaul operations.
- Easy to use—just score and remove tube tips, insert special five-tube holder into manifold, and connect manifold to pump
- After 10 complete pump strokes, tubes are ready for evaluation
Order Info
Individual test sets and adapter also sold separately.
†Each Simultest Set consists of five detector tubes in a rubber holder—ready for use. Each test set can be completed in one minute or less.
Specifications
Hazmat Simultaneous Test Kit | |
Accuro™ pump, pump spare parts kit, air currents kit, extension hose with adapter, test set opener, deluxe tube opener, laminated instruction cards, simultaneous test set I, simultaneous test set II, simultaneous test set III, hard Plast |