missing translation for 'onlineSavingsMsg'
Learn More

DPCD Monoclonal Antibody (K1E042_4B5), Invitrogen™

Mouse Monoclonal Antibody

$1557.73 - $2745.56

Specifications

Antigen DPCD
Clone K1E042_4B5
Concentration 1 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MA556055
View Documents
Invitrogen™
MA556055
200 μg
Each for $2,745.56
Only null left
 
MA555675
View Documents
Invitrogen™
MA555675
50 μg
Each for $1,557.73
Only null left
 
Description

Description

Sequence of this protein is as follows: RTAQKTALLQ DGRRKVHYLF PDGKEMAEEY DEKTSELLVR KWRVKSALGA MGQWQLEVGD PAPLGAGNLG PELIKESNAN PIFMRKDTKM SFQWRIRNLP YPKDVYSVSV DQKERCIIVR TTNKKYYKKF SIPDLDRHQL PLDDALLSFA HANCTLIISY QKPKEVVVAE SELQKELKKV KTAHSNDGDC

DPCD Monoclonal antibody specifically detects DPCD in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot
Specifications

Specifications

DPCD
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide
5330431N19Rik; deleted in a mouse model of primary ciliary dyskinesia; deleted in primary ciliary dyskinesia; deleted in primary ciliary dyskinesia homolog (mouse); DPCD; Ndac; neighbor of Dac; protein DPCD; RGD1307648; RP11-529I10.4; RP11-529I104
Recombinant human DPCD (immunogen range is 11-200).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
DPCD
K1E042_4B5
Immunoprecipitation, Western Blot
Unconjugated
Mouse
Human, Mouse
Q8BPA8, Q9BVM2
226162, 25911
IgG2b
Protein A
Antibody
Videos
SDS
Documents

Documents