missing translation for 'onlineSavingsMsg'
Learn More

Doxorubicin Hydrochloride 95.0+%, TCI America™

$104.38 - $260.97

Chemical Identifiers

CAS 25316-40-9
Molecular Formula C27H30ClNO11
Molecular Weight (g/mol) 579.98
MDL Number MFCD00077757,MFCD00077757,MFCD00941448
InChI Key MWWSFMDVAYGXBV-FGBJBKNOSA-N
Synonym doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
PubChem CID 129626538
IUPAC Name hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
SMILES [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D419325MG
View Documents
TCI America
D419325MG
25 mg
Each for $104.38
Only null left
 
D4193100MG
View Documents
TCI America
D4193100MG
100 mg
Each for $260.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

25316-40-9
579.98
MWWSFMDVAYGXBV-FGBJBKNOSA-N
129626538
[H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O
C27H30ClNO11
MFCD00077757,MFCD00077757,MFCD00941448
doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride

Specifications

25316-40-9
MFCD00077757,MFCD00077757,MFCD00941448
doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
[H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O
579.98
579.98
Crystalline Powder
C27H30ClNO11
25 mg
MWWSFMDVAYGXBV-FGBJBKNOSA-N
hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
129626538
≥95.0% (HPLC)
Doxorubicin Hydrochloride
Videos
Safety and Handling

Safety and Handling

RTECSNumber : QI9295900

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'