Learn More
Thermo Scientific Chemicals DL-Tryptophan, 99%
DL-Tryptophan, CAS # 54-12-6, is an α-amino acid that is commonly used in the synthesis of proteins. It is an essential amino acid precursor for the formation of serotonin, melatonin, and vitamin B3. | CAS: 54-12-6 | C11H12N2O2 | 204.23 g/mol
$52.25 - $333.64
Chemical Identifiers
CAS | 54-12-6 |
---|---|
Molecular Formula | C11H12N2O2 |
Molecular Weight (g/mol) | 204.23 |
MDL Number | MFCD00064339 |
InChI Key | QIVBCDIJIAJPQS-UHFFFAOYSA-N |
Synonym | dl-tryptophan, 2-amino-3-1h-indol-3-yl propanoic acid, racemic tryptophan, dl-trytophane, dl-trytophan, +--tryptophan, h-dl-trp-oh, dl-3beta-indolylalanine, dl-tryptophane, tryptophan . |
PubChem CID | 1148 |
ChEBI | CHEBI:27897 |
IUPAC Name | 2-amino-3-(1H-indol-3-yl)propanoic acid |
SMILES | NC(CC1=CNC2=CC=CC=C12)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0593606
|
Thermo Scientific Chemicals
L0593606 |
5 g |
Each for $52.25
|
|
|||||
AAL0593614
|
Thermo Scientific Chemicals
L0593614 |
25 g |
Each for $119.52
|
|
|||||
AAL0593622
|
Thermo Scientific Chemicals
L0593622 |
100 g |
Each for $333.64
|
|
|||||
Description
DL-Tryptophan is an amino acid precursor of serotonin and melatonin. It is a dietary supplement for use as an antidepressant, anxiolytic, and sleep aid. DL-tryptophan also can be used as feed nutrition enhancer, antioxidants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• DL-Tryptophan is an α-amino acid, and an essential amino acid precursor of serotonin, melatonin, and vitamin B3
Application
• DL-Tryptophan shows antioxidant activity• It has a role as a Daphnia magna metabolite
• It has been explored in in vitro studies to evaluate the antimicrobial and immunoregulatory functions on indoleamine 2,3-dioxygenase
• It has been used as a medium supplement in Psilocybe atrobrunnea and Psilocybe serbica for the synthesis of compounds
• It can be used as a medium supplement for culturing yeast
Chemical Identifiers
54-12-6 | |
204.23 | |
QIVBCDIJIAJPQS-UHFFFAOYSA-N | |
1148 | |
2-amino-3-(1H-indol-3-yl)propanoic acid |
C11H12N2O2 | |
MFCD00064339 | |
dl-tryptophan, 2-amino-3-1h-indol-3-yl propanoic acid, racemic tryptophan, dl-trytophane, dl-trytophan, +--tryptophan, h-dl-trp-oh, dl-3beta-indolylalanine, dl-tryptophane, tryptophan . | |
CHEBI:27897 | |
NC(CC1=CNC2=CC=CC=C12)C(O)=O |
Specifications
∽280°C (decomposition) | |
C11H12N2O2 | |
86196 | |
Soluble I water. 10g/L (20°C),Hcl and alkalies. Sparingly soluble in alcohol. | |
NC(CC1=CNC2=CC=CC=C12)C(O)=O | |
5 g | |
1148 | |
204.23 | |
DL-Tryptophan |
54-12-6 | |
MFCD00064339 | |
dl-tryptophan, 2-amino-3-1h-indol-3-yl propanoic acid, racemic tryptophan, dl-trytophane, dl-trytophan, +--tryptophan, h-dl-trp-oh, dl-3beta-indolylalanine, dl-tryptophane, tryptophan . | |
QIVBCDIJIAJPQS-UHFFFAOYSA-N | |
2-amino-3-(1H-indol-3-yl)propanoic acid | |
204.23 | |
CHEBI:27897 | |
99% |
Safety and Handling
EINECSNumber : 200-194-9
RTECSNumber : YN6129200
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only