missing translation for 'onlineSavingsMsg'
Learn More

DL-Tropic Acid 98.0+%, TCI America™

$113.71 - $564.81

Chemical Identifiers

CAS 552-63-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004255
InChI Key JACRWUWPXAESPB-UHFFFAOYNA-N
Synonym tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid
PubChem CID 10726
ChEBI CHEBI:30765
IUPAC Name 3-hydroxy-2-phenylpropanoic acid
SMILES OCC(C(O)=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T053325G
View Documents
TCI America
T053325G
25 g
Each for $113.71
Only null left
 
T0533250G
View Documents
TCI America
T0533250G
250 g
Each for $564.81
Only null left
 
Description

Description

Specifications

Chemical Identifiers

552-63-6
166.18
JACRWUWPXAESPB-UHFFFAOYNA-N
10726
3-hydroxy-2-phenylpropanoic acid
C9H10O3
MFCD00004255
tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid
CHEBI:30765
OCC(C(O)=O)C1=CC=CC=C1

Specifications

552-63-6
White
MFCD00004255
tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid
OCC(C(O)=O)C1=CC=CC=C1
166.18
CHEBI:30765
≥98.0% (GC,T)
DL-Tropic Acid
118°C
C9H10O3
25 g
JACRWUWPXAESPB-UHFFFAOYNA-N
3-hydroxy-2-phenylpropanoic acid
10726
166.18
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3083

RTECSNumber : CY8535000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'