missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Mandelic Acid, 98%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor MA16012KGBL
Specifications
611-72-3 | |
98% | |
12 kg |
1 | |
Reagent | |
Poly Pail |
Chemical Identifiers
611-72-3 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.