missing translation for 'onlineSavingsMsg'
Learn More

DL-Mandelic Acid, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor MA16025KG

 View more versions of this product

Catalog No. 18610298


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

611-72-3
98%
2.5 kg
1
Reagent
Poly Pail

Chemical Identifiers

611-72-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.