missing translation for 'onlineSavingsMsg'
Learn More

DL-Malic Acid 99.0+%, TCI America™

$46.60 - $59.65

Chemical Identifiers

CAS 6915-15-7
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.087
MDL Number MFCD00064212
InChI Key BJEPYKJPYRNKOW-UHFFFAOYSA-N
Synonym malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid, butanedioic acid, hydroxy, kyselina jablecna, pomalus acid, hydroxybutanedioic acid, deoxytetraric acid
PubChem CID 525
ChEBI CHEBI:6650
IUPAC Name 2-hydroxybutanedioic acid
SMILES C(C(C(=O)O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M002025G
View Documents
TCI America
M002025G
25 g
Each for $46.60
Only null left
 
M0020500G
View Documents
TCI America
M0020500G
500 g
Each for $59.65
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6915-15-7
134.087
BJEPYKJPYRNKOW-UHFFFAOYSA-N
525
2-hydroxybutanedioic acid
C4H6O5
MFCD00064212
malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid, butanedioic acid, hydroxy, kyselina jablecna, pomalus acid, hydroxybutanedioic acid, deoxytetraric acid
CHEBI:6650
C(C(C(=O)O)O)C(=O)O

Specifications

130°C
White
MFCD00064212
BJEPYKJPYRNKOW-UHFFFAOYSA-N
2-hydroxybutanedioic acid
525
134.09
25 g
DL-Malic Acid
6915-15-7
C4H6O5
malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid, butanedioic acid, hydroxy, kyselina jablecna, pomalus acid, hydroxybutanedioic acid, deoxytetraric acid
C(C(C(=O)O)O)C(=O)O
134.087
CHEBI:6650
≥99.0% (T)
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1442

RTECSNumber : ON7175000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'