Learn More
Thermo Scientific Chemicals DL-Lysine monohydrochloride, 99%
CAS: 70-53-1 | C6H15ClN2O2 | 182.65 g/mol
$68.85 - $181.82
Chemical Identifiers
CAS | 70-53-1 |
---|---|
Molecular Formula | C6H15ClN2O2 |
Molecular Weight (g/mol) | 182.65 |
MDL Number | MFCD00064563 |
InChI Key | BVHLGVCQOALMSV-UHFFFAOYSA-N |
Synonym | dl-lysine monohydrochloride, 2,6-diaminohexanoic acid hydrochloride, dl-lysine hydrochloride, lysine monohydrochloride, dl-lysine mono hydrochloride, lysine, hydrochloride 1:1, lysine, monohydrochloride, dl-2,6-diaminohexanoic acid monohydrochloride, lysine, dl, dl-lysinemonohydrochloride |
PubChem CID | 12047 |
IUPAC Name | 2,6-diaminohexanoic acid;hydrochloride |
SMILES | Cl.NCCCCC(N)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1106614
|
Thermo Scientific Chemicals
A1106614 |
25 g |
Each for $68.85
|
|
|||||
AAA1106622
|
Thermo Scientific Chemicals
A1106622 |
100 g |
Each for $181.82
|
|
|||||
Description
DL-Lysine monohydrochloride acts as an intermediate in the preparation of 3-amino-azepan-2-one. It also serves as an essential amino acid. It is used as animal feed. In addition to this, it is used in the construction of polylysine polymers, which finds application in lysine-based polypeptide gels and films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Lysine monohydrochloride acts as an intermediate in the preparation of 3-amino-azepan-2-one. It also serves as an essential amino acid. It is used as animal feed. In addition to this, it is used in the construction of polylysine polymers, which finds application in lysine-based polypeptide gels and films.
Solubility
Soluble in water. Insoluble in ethanol, ethyl ether, acetone, benzene and common neutral solvents.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
70-53-1 | |
182.65 | |
BVHLGVCQOALMSV-UHFFFAOYSA-N | |
12047 | |
Cl.NCCCCC(N)C(O)=O |
C6H15ClN2O2 | |
MFCD00064563 | |
dl-lysine monohydrochloride, 2,6-diaminohexanoic acid hydrochloride, dl-lysine hydrochloride, lysine monohydrochloride, dl-lysine mono hydrochloride, lysine, hydrochloride 1:1, lysine, monohydrochloride, dl-2,6-diaminohexanoic acid monohydrochloride, lysine, dl, dl-lysinemonohydrochloride | |
2,6-diaminohexanoic acid;hydrochloride |
Specifications
267°C (decomposition) | |
C6H15ClN2O2 | |
4711993 | |
Soluble in water. Insoluble in ethanol,ethyl ether,acetone,benzene and common neutral solvents. | |
Cl.NCCCCC(N)C(O)=O | |
25 g | |
12047 | |
99% |
70-53-1 | |
MFCD00064563 | |
dl-lysine monohydrochloride, 2,6-diaminohexanoic acid hydrochloride, dl-lysine hydrochloride, lysine monohydrochloride, dl-lysine mono hydrochloride, lysine, hydrochloride 1:1, lysine, monohydrochloride, dl-2,6-diaminohexanoic acid monohydrochloride, lysine, dl, dl-lysinemonohydrochloride | |
BVHLGVCQOALMSV-UHFFFAOYSA-N | |
2,6-diaminohexanoic acid;hydrochloride | |
182.65 | |
182.65 | |
DL-Lysine monohydrochloride |
Safety and Handling
EINECSNumber : 200-739-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only