missing translation for 'onlineSavingsMsg'
Learn More

DL-Dithiothreitol 98.0+%, TCI America™

$98.79 - $296.38

Chemical Identifiers

CAS 3483-12-3
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00004877
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
PubChem CID 446094
ChEBI CHEBI:42170
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
SMILES C(C(C(CS)O)O)S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D36471G
View Documents
TCI America
D36471G
1 g
Each for $98.79
Only null left
 
D36475G
View Documents
TCI America
D36475G
5 g
Each for $296.38
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3483-12-3
154.24
VHJLVAABSRFDPM-IMJSIDKUSA-N
446094
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
C4H10O2S2
MFCD00004877
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
CHEBI:42170
C(C(C(CS)O)O)S

Specifications

3483-12-3
White
C4H10O2S2
1 g
VHJLVAABSRFDPM-IMJSIDKUSA-N
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
446094
154.24
Crystalline Powder
43°C
130°C
MFCD00004877
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
C(C(C(CS)O)O)S
154.24
CHEBI:42170
≥98.0% (T)
DL-Dithiothreitol [for Electrophoresis]
Videos
Safety and Handling

Safety and Handling

RTECSNumber : XO8576500

TSCA : Yes

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'