missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Alanine, 99%

CAS: 302-72-7 | C3H7NO2 | 89.09 g/mol

$71.14 - $619.62

Chemical Identifiers

CAS 302-72-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064408
InChI Key QNAYBMKLOCPYGJ-UHFFFAOYNA-N
Synonym dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine
PubChem CID 602
ChEBI CHEBI:16449
IUPAC Name 2-aminopropanoic acid
SMILES CC(N)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1223030
View Documents
Thermo Scientific Chemicals
A1223030
250 g
Each for $71.14
Only null left
 
AAA122300B
View Documents
Thermo Scientific Chemicals
A122300B
1000 g
Each for $166.66
Only null left
 
AAA122300I
View Documents
Thermo Scientific Chemicals
A122300I
5000 g
Each for $619.62
Only null left
 
Description

Description

DL-Alanine is used in mixed ligand (chelate) studies with transition metals such as Cu(II), Zn(ll) and Cd(ll). It is a functional component in the alanine cycle which regulates the level of alanine aminotransferases which can be used as a diagnostic tool to predict type II diabetes. It is used as a model reagent for development of amino acid racemic resolution techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Alanine is used in mixed ligand (chelate) studies with transition metals such as Cu(II), Zn(ll) and Cd(ll). It is a functional component in the alanine cycle which regulates the level of alanine aminotransferases which can be used as a diagnostic tool to predict type II diabetes. It is used as a model reagent for development of amino acid racemic resolution techniques.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

302-72-7
89.09
QNAYBMKLOCPYGJ-UHFFFAOYNA-N
602
2-aminopropanoic acid
C3H7NO2
MFCD00064408
dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine
CHEBI:16449
CC(N)C(O)=O

Specifications

∼292°C (decomposition)
C3H7NO2
635807
dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine
QNAYBMKLOCPYGJ-UHFFFAOYNA-N
1.42
250 g
602
89.09
DL-Alanine
302-72-7
MFCD00064408
14,204
Soluble in water. Slightly soluble in alcohol.
CC(N)C(O)=O
2-aminopropanoic acid
89.09
CHEBI:16449
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 206-126-4

RTECSNumber : AY2980000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only