missing translation for 'onlineSavingsMsg'
Learn More

FEATHER Dissecting Blade

FEATHER Dissecting Blades are designed for autopsy. The extra-thick stainless steel blade provides high rigidity and extra-long life. Can be used with autoclavable handle designed for easy blade replacement and cleaning.

$78.99 - $81.86

Specifications

For Use With (Equipment) FEATHER Dissecting Blade Handle F-60
Length (Metric) 77 mm
Material Stainless Steel
Quantity 20 Pk.
Width (Metric) 0.6 mm
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
08-924-25B
Accessories View Documents
FEATHER
61
No. 61 (Wide blade)
Pack of 20 Each for $81.86
Only null left
 
08-924-25C
Accessories View Documents
FEATHER
62
No. 62 (Narrow blade)
Pack of 20 Each for $78.99
Only null left
 
Description

Description

  • Made in Japan
Specifications

Specifications

FEATHER Dissecting Blade Handle F-60
Stainless Steel
0.6 mm
77 mm
20 Pk.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'