Learn More
Epredia™ Disposable Base Molds
Use Epredia™ Disposable Base Molds with all standard-size cassettes.
Supplier:  Epredia™ 58953
 View more versions of this product
Catalog No. 36-101-4339
Only null left
Explore available promotions
Description
Description
  • Cleaning associated with metal molds is not required
  • Transparency facilitates tissue orientation
Ordering Information:
  • Sold in cases of four boxes.
Specifications
Specifications
Base Mold
Plastic
37 x 24 mm
Single-use
Product Suggestions
Product Suggestions