missing translation for 'onlineSavingsMsg'
Learn More

Dynalon™ Dispensing Bottles/Screw-on Caps

$79.20 - $161.50

Specifications

Color Translucent, White
For Use With (Application) Dispensing
Includes Dropper cap
Material LDPE
Sterility Non-sterile
View More Specs

Products 4
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S31720
View Documents
Dynalon™
BGE514
30 mL
Pack of 12 Each for $81.15
Case of 4 Pack for $293.25
Only null left
 
S31722
View Documents
Dynalon™
BGE520
60 mL
Pack of 12 Each for $79.20
Case of 3 Pack for $215.85
Only null left
 
S31723
View Documents
Dynalon™
BGE528
125 mL
Pack of 12 Each for $102.10
Case of 3 Pack for $277.95
Only null left
 
S31724
View Documents
Dynalon™
BGE530
250 mL
Pack of 12 Each for $161.50
Case of 2 Pack for $318.35
Only null left
 
Description

Description

Low-density polyethylene dispensing bottles feature screw-on caps and captive covers which cannot be lost or accidentally placed on the wrong bottle.

  • Ideal for dispensing drop by drop or in a continuous stream
  • Controlled by squeezing the bottle

Specifications

Specifications

Translucent, White
Dropper cap
Non-sterile
Dispensing Bottle
Dispensing
LDPE
Not Autoclavable
Dropper Cap
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'