missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylamine, ACS Reagent Grade, Ricca Chemical
$153.38 - $153.38
Chemical Identifiers
CAS | 122-39-4 |
---|---|
Molecular Formula | C12H11N |
Molecular Weight (g/mol) | 169.227 |
InChI Key | DMBHHRLKUKUOEG-UHFFFAOYSA-N |
Synonym | diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald |
PubChem CID | 11487 |
ChEBI | CHEBI:4640 |
IUPAC Name | N-phenylaniline |
SMILES | C1=CC=C(C=C1)NC2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
RDCD0650100
|
Ricca Chemical Company
RDCD0650100B1 |
100 g | White Poly Wide Mouth Bottle |
Each for $153.38
|
|
||||
Chemical Identifiers
122-39-4 | |
169.227 | |
diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald | |
CHEBI:4640 | |
C1=CC=C(C=C1)NC2=CC=CC=C2 |
C12H11N | |
DMBHHRLKUKUOEG-UHFFFAOYSA-N | |
11487 | |
N-phenylaniline |
Specifications
122-39-4 | |
100.0 | |
White Poly Wide Mouth Bottle | |
100 g | |
DMBHHRLKUKUOEG-UHFFFAOYSA-N | |
N-phenylaniline | |
11487 | |
ACS | |
Diphenylamine |
98.0 | |
302°C | |
C12H11N | |
diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald | |
C1=CC=C(C=C1)NC2=CC=CC=C2 | |
169.227 | |
CHEBI:4640 | |
Crystalline Powder or Lumps |
Safety and Handling
ShelfLife : 60 months