missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl Disulfide 99.0+%, TCI America™
$49.13 - $277.79
Chemical Identifiers
CAS | 882-33-7 |
---|---|
Molecular Formula | C12H10S2 |
Molecular Weight (g/mol) | 218.332 |
MDL Number | MFCD00003065 |
InChI Key | GUUVPOWQJOLRAS-UHFFFAOYSA-N |
Synonym | diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene |
PubChem CID | 13436 |
IUPAC Name | (phenyldisulfanyl)benzene |
SMILES | C1=CC=C(C=C1)SSC2=CC=CC=C2 |
Chemical Identifiers
882-33-7 | |
218.332 | |
GUUVPOWQJOLRAS-UHFFFAOYSA-N | |
13436 | |
C1=CC=C(C=C1)SSC2=CC=CC=C2 |
C12H10S2 | |
MFCD00003065 | |
diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene | |
(phenyldisulfanyl)benzene |
Specifications
882-33-7 | |
White-Yellow | |
MFCD00003065 | |
3077 | |
GUUVPOWQJOLRAS-UHFFFAOYSA-N | |
(phenyldisulfanyl)benzene | |
13436 | |
≥99.0% (GC) | |
Diphenyl Disulfide |
61°C | |
C12H10S2 | |
25 g | |
diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene | |
C1=CC=C(C=C1)SSC2=CC=CC=C2 | |
218.332 | |
218.33 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1124
RTECSNumber : SS6825000
TSCA : Yes