missing translation for 'onlineSavingsMsg'
Learn More

Dimidium Bromide, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor D2499100GMBL

 View more versions of this product

Catalog No. 18610252

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

518-67-2
Amber Glass Bottle
0.97
1
100 g
Reagent

Chemical Identifiers

518-67-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For use by qualified personnel for research and development only. Read and understand the label and Safety Data Sheet (SDS) prior to use.