missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylamine, 26% w/v, Reagents
Dimethylamine, 26% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$363.73 - $1357.67
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.02 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | N-Methylmethanamine |
IUPAC Name | water |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-335-416
|
Labchem Inc
CD2611001C |
1 L |
Each for $363.73
|
|
|||||
01-335-415
|
Labchem Inc
CD2611004C1 |
4 L |
Each for $1,357.67
|
|
|||||
Chemical Identifiers
7732-18-5 | |
18.02 | |
N-Methylmethanamine | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
7732-18-5,124-40-3 | |
10 | |
H2O | |
0.9 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
26% | |
Liquid |
Colorless to Light Yellow | |
Glass Bottle | |
1 L | |
N-Methylmethanamine | |
O | |
18.02 | |
Reagent | |
Dimethylamine, 26% w/v |