Learn More

Chemical Identifiers
CAS | 67-68-5 |
---|---|
Molecular Formula | C2H6OS |
Molecular Weight (g/mol) | 78.13 |
MDL Number | MFCD00002089 |
InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide |
PubChem CID | 679 |
ChEBI | CHEBI:28262 |
IUPAC Name | methanesulfinylmethane |
SMILES | CS(C)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
D1594
|
Thermo Fisher Scientific
D1594 |
4 L | Amber Glass |
Each for $1,295.23
Case of 4 Each for $5,180.92
|
|
||||
Chemical Identifiers
67-68-5 | |
78.13 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
679 | |
methanesulfinylmethane |
C2H6OS | |
MFCD00002089 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CHEBI:28262 | |
CS(C)=O |
Specifications
18.4°C | |
0.01 AU max., 0.02 AU max. at 335nm, 0.02 AU max. at 330nm, 0.06 AU max. at 310nm, 0.1 AU max. at 300nm, 0.25 AU max. at 280nm, 0.2 AU max. at 290nm, 0.3 AU max. at 275nm, 0.4 AU max. at 270nm, 1.00 AU max. at 263nm | |
>190°C | |
189°C | |
87°C (188.6°F) | |
Amber Glass | |
1.4770 to 1.4800 (at 25°C) | |
99.8% min by GC | |
C2H6OS | |
MFCD00002089 | |
1.98 mPa/s at 25°C | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CS(C)=O | |
78.13 | |
CHEBI:28262 | |
≥99.9% | |
Dimethyl Sulfoxide |
67-68-5 | |
Colorless | |
1.100g/cm³ | |
4 L | |
Pass Test | |
Liquid | |
0.005% Max | |
0.001mEq/g max. | |
(CH3)2SO | |
0.55mbar at 20°C | |
0.01% max. | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
methanesulfinylmethane | |
679 | |
78.13 | |
Mobile phase for HPLC applications |
Safety and Handling

WARNING!
Emergency Overview
Combustible liquid. DMSO readily penetrates skin and may carry other dissolved chemicals into the body. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.
NFPA
Health:2
Flammability:2
Instability:0