missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl sulfoxide, ≥99%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219605580
Specifications
Dimethyl sulfoxide | |
100 mL | |
≥99% | |
MFCD00002089 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
methylsulfinylmethane | |
679 | |
78.1 |
67-68-5 | |
Liquid | |
C2H6OS | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CS(C)=O | |
78.13 | |
CHEBI:28262 | |
≥99% |
Chemical Identifiers
67-68-5 | |
78.13 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
679 | |
CS(C)=O |
C2H6OS | |
MFCD00002089 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CHEBI:28262 |