missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl bromomalonate, 90%, tech.

CAS: 868-26-8 | C5H7BrO4 | 211.02 g/mol

$350.45 - $350.45

Chemical Identifiers

CAS 868-26-8
Molecular Formula C5H7BrO4
Molecular Weight (g/mol) 211.02
MDL Number MFCD00025865
InChI Key NEMOJKROKMMQBQ-UHFFFAOYSA-N
Synonym dimethyl bromomalonate, dimethyl 2-bromomalonate, dimethylbromomalonate, propanedioic acid, bromo-, dimethyl ester, 2-bromo dimethyl malonate, 1,3-dimethyl 2-bromopropanedioate, dimethylbromomalo-nate, acmc-209qbk, dimethyl-2-bromomalonate, dimethyl 2-bromomalonate #
PubChem CID 70087
IUPAC Name dimethyl 2-bromopropanedioate
SMILES COC(=O)C(C(=O)OC)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC305190250
SDS View Documents
Thermo Scientific Chemicals
305190250
25 mL Glass bottle
Each for $350.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

868-26-8
211.02
NEMOJKROKMMQBQ-UHFFFAOYSA-N
70087
COC(=O)C(C(=O)OC)Br
C5H7BrO4
MFCD00025865
dimethyl bromomalonate, dimethyl 2-bromomalonate, dimethylbromomalonate, propanedioic acid, bromo-, dimethyl ester, 2-bromo dimethyl malonate, 1,3-dimethyl 2-bromopropanedioate, dimethylbromomalo-nate, acmc-209qbk, dimethyl-2-bromomalonate, dimethyl 2-bromomalonate #
dimethyl 2-bromopropanedioate

Specifications

868-26-8
1.6000g/mL
>110°C
88% min. (GC)
C5H7BrO4
CH3O2CCH(Br)CO2CH3
25 mL
dimethyl bromomalonate, dimethyl 2-bromomalonate, dimethylbromomalonate, propanedioic acid, bromo-, dimethyl ester, 2-bromo dimethyl malonate, 1,3-dimethyl 2-bromopropanedioate, dimethylbromomalo-nate, acmc-209qbk, dimethyl-2-bromomalonate, dimethyl 2-bromomalonate #
COC(=O)C(C(=O)OC)Br
211.02
211.02
Technical
Dimethyl bromomalonate, tech., 90%
Colorless to Yellow
105°C to 108°C (11.0 mmHg)
Authentic
Glass bottle
1.4610 to 1.465
MFCD00025865
1.6
NEMOJKROKMMQBQ-UHFFFAOYSA-N
dimethyl 2-bromopropanedioate
70087
90%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 212-775-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only