missing translation for 'onlineSavingsMsg'
Learn More

Diisopropanolamine, Spectrum™ Chemical

$256.79 - $4560.65

Chemical Identifiers

CAS 110-97-4
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610204
View Documents
Spectrum Chemical Mfg Cor
D1324500GM
500 g Amber Glass Bottle
Each for $256.79
Only null left
 
18610203
View Documents
Spectrum Chemical Mfg Cor
D132440LBBL
40 lb. Poly Pail
Each for $4,560.65
Only null left
 
18610202
View Documents
Spectrum Chemical Mfg Cor
D132425KG
2.5 kg Poly Pail
Each for $838.88
Only null left
 
Description

Description

Specifications

Chemical Identifiers

110-97-4

Specifications

110-97-4
Amber Glass Bottle
Reagent
1
500 g
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.