missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylenetriaminepentaacetic Acid, 98%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor D232325KG
Specifications
67-43-6 | |
Poly Bottle | |
0.98 |
1 | |
2.5 kg | |
Reagent |
Chemical Identifiers
67-43-6 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.