missing translation for 'onlineSavingsMsg'
Learn More

Diethanolamine, Reagent, ACS, 98.5%, Spectrum™ Chemical

D1033, 111-42-2, C4H11NO2

Supplier:  Spectrum Chemical Mfg Cor D1033500MLGL

 View more versions of this product

Catalog No. 18602184


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Diethanolamine, Reagent, ACS is also referred to as DEA, is an organic compound that in its pure form is a white solid but it is most often encountered as a clear liquid that has a viscous texture. It is a secondary amine and diol and is water soluble. It is used as a corrosion inhibitor and surfactant that is used in the oil refining industry to help remove hydrogen sulfide from sour gas. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.
Specifications

Specifications

111-42-2
Amber Glass Bottle
MFCD00002843
ZBCBWPMODOFKDW-UHFFFAOYSA-N
2-[(2-hydroxyethyl)amino]ethan-1-ol
98.5%
100%
C4H11NO2
500 mL
OCCNCCO
105.14
ACS

Chemical Identifiers

111-42-2
105.14
ZBCBWPMODOFKDW-UHFFFAOYSA-N
OCCNCCO
C4H11NO2
MFCD00002843
2-[(2-hydroxyethyl)amino]ethan-1-ol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions