missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Didodecyl Sulfoxide 98.0+%, TCI America™
$313.16 - $999.13
Chemical Identifiers
CAS | 2168-96-9 |
---|---|
Molecular Formula | C24H50OS |
Molecular Weight (g/mol) | 386.72 |
MDL Number | MFCD00039865 |
InChI Key | PZKDFFVFMXTDIP-UHFFFAOYSA-N |
Synonym | Dodecyl Sulfoxide |
PubChem CID | 75109 |
IUPAC Name | 1-(dodecane-1-sulfinyl)dodecane |
SMILES | CCCCCCCCCCCCS(=O)CCCCCCCCCCCC |
Chemical Identifiers
2168-96-9 | |
386.72 | |
PZKDFFVFMXTDIP-UHFFFAOYSA-N | |
75109 | |
CCCCCCCCCCCCS(=O)CCCCCCCCCCCC |
C24H50OS | |
MFCD00039865 | |
Dodecyl Sulfoxide | |
1-(dodecane-1-sulfinyl)dodecane |
Specifications
2168-96-9 | |
White-Yellow | |
MFCD00039865 | |
Dodecyl Sulfoxide | |
CCCCCCCCCCCCS(=O)CCCCCCCCCCCC | |
386.72 | |
386.72 | |
Crystalline Powder |
91°C | |
C24H50OS | |
5 g | |
PZKDFFVFMXTDIP-UHFFFAOYSA-N | |
1-(dodecane-1-sulfinyl)dodecane | |
75109 | |
≥98.0% (HPLC) | |
Didodecyl Sulfoxide |
Safety and Handling
TSCA : No